Safety Shields: Remain Splash-Free Using Shields and Other Safety Devices
Safety Shields
Cole-Parmer® Benchtop Splash-Guard
Working in a lab with biohazards, chemicals, or other potentially dangerous substances? Protect yourself from splashes and flying debris with splash shields and safety shields. Some shields even protect you from beta radiation.
Cole-Parmer® Benchtop Splash-Guard
Cole-Parmer® Benchtop Splash-Guard
Because shields are crafted of clear materials, such as Plexiglas® acrylic, and polycarbonate, they do not limit your vision or prevent you from accessing the bench. Most shields adjust to provide the flexibility you need for a variety of projects.
Shield options include freestanding, magnetically clamped, and adjustable neck clamp versions. For greater stability, look for weighted shields with a solid base to hold the shield in place.
Twin bracket clamp models provide the strength and stability to hold larger and heavier shields without compromising adjustability.
When selecting a shield, make sure that it is designed to protect you from the type of hazard you will encounter, such as biohazards and beta radiation.
See our full selection of safety shields.
Be the first to comment on "Safety Shields: Remain Splash-Free Using Shields and Other Safety Devices"