SARS-CoV-2 Omicron External Run Control Now Available
ZeptoMetrix SARS-CoV-2 Omicron Variant Molecular Quality Control
Ideal reference material for molecular diagnostic testing applications.
If you need quality controls for the SARS-CoV-2 Omicron variant, you will be interested to know that the new ZeptoMetrix® SARS-CoV-2 Omicron External Run Control has been added to the company’s expansive line of infectious disease quality controls and is now available for pre-order. SARS-CoV-2 control is a significant part of the testing process—ZeptoMetrix External Run Controls and Verification Panels on a consistent basis helps you to gain additional confidence in test results.
Reliable testing
ZeptoMetrix continues to lead the fight against Omicron as a partner to its customers. The company’s scientists monitor SARS-CoV-2 mutations and move quickly to offer the latest quality controls, so you can provide the most reliable testing for your customers. ZeptoMetrix consistently works to bring you not only quality controls for SARS-CoV-2 but also quality controls for many infectious diseases.
As with this latest Omicron control, all ZeptoMetrix NATtrol™ quality controls undergo a process that renders highly purified microorganisms non-infectious while allowing the internal nucleic acids to remain intact. The resulting product is an ideal reference material for molecular diagnostic testing applications. Representing a total process control, from DNA extraction through amplification and eventual detection, NATtrol products are ready to use, refrigerator stable, and safe for operators to handle. The routine and repetitive use of NATtrol external run controls and verification panels helps scientists monitor performance and identify trends in daily test variations, test kit lot changes, and variations among individual operators.
For more information on SARS-CoV-2 Omicron control, visit www.zeptometrix.com.
Related Article
[Webinar On-Demand] Reduce Errors and Increase Confidence in Testing with Quality Controls
Sign up for the blog banner
Be the first to comment on "SARS-CoV-2 Omicron External Run Control Now Available"