Save Time with Multiposition Stirring Hot Plates
multi-position stirring hot plate
Cole-Parmer® Nine-Position Stirring Hot Plate
Whenever you can capture lost time it makes your job more efficient. That’s why Multiposition Stirring Hot Plates are worthwhile for your lab. You can stir or heat multiple samples at the same time—no need to wait until one is finished to start the next one.
Because each stirring position is individually controlled, you also can perform different mixing applications at the same time. These hot plates can heat only, stir only, or heat and stir at the same time to maximize versatility.
From a safety angle, the controls are designed so that you don’t have to touch the hot plate to make adjustments.
Some multiposition units are programmable to work unattended while you are busy elsewhere in the lab. They allow you to set the temperature, stirring speed, time, and heating ramp rate.
Similar to the multiposition stirring hot plate is the multiposition extraction heater/stirrers which allow liquids with different boiling points to be heated or stirred next to each other.
View our selection of stirring hot plates. For questions about which is best for you, contact our Technical Support Experts.
Be the first to comment on "Save Time with Multiposition Stirring Hot Plates"