Extract DNA from plant and animal tissue quickly
If you are working with DNA extraction and would like fast and seamless isolation of DNA from a wide variety of animal tissues and cells, take a look at the new Spex Genomic DNA Isolation Kit. These kits are optimized to work with the Genomax™ and allows for fast and seamless isolation of DNA from a wide variety of animal tissues and cells, including mammalian, poultry, and fish, and soft and tough plant tissue.
The kit combines the use of chaotropic salts and detergents to disrupt animal tissues or cells permitting the selective binding of the DNA to a silica column. Proteinase K and RNase A are then added to implement DNA isolation. An ethanol wash buffer is used to remove any additional elements or contamination from the silica column. The column is dried by centrifugation and DNA is eluted using the elution buffer.
Benefits
- Faster process and 1-touch methods saves you time
- More samples per kit
- Lower cost saves you money
- Up to 85% higher yields and higher quality
Watch Video
Order your free sample kit!
Be the first to comment on "[Video] How to Simplify DNA Extraction"