A professional researcher working in academia told us the Digi-Sense® Calibrated Fridge/Freezer Digital Thermometer is an “Excellent device for staying in compliance by monitoring your cooling and freezer compartments.”
What’s so special about this thermometer? With a triple display, it shows high, low, and current temperature simultaneously. It allows you to easily monitor temperature in liquids, baths, air/gas, incubators, refrigerators, or freezers. You can keep track of temperature conditions overnight or for any time period—minimum and maximum temperatures are stored until the memory is cleared.
You can also set high and low alarms in 1° increments. The alarm sounds when the temperature rises above or falls below set points, and continues to sound even if temperature returns to nonalarm range. All in all, it is “accurate, intuitive, and useful,” as another customer pointed out.
Find out more about this Fridge/Freezer Digital Thermometer. See our full line of digital thermometers, especially for the food industry.
Be the first to comment on "Stay in Compliance with Refrigerator/Freezer Thermometer"