Take Three: Timely New Technologies from Thermo Scientific
thermo scientific low temperature incubator
Packed with efficiencies, the three innovations below all enhance your lab by delivering reliable functionality while also saving you precious space.
Thermo-Scientific-Low-Temperature-IncubatorThermo Scientific Low Temperature Incubator meets American Public Health Association (APHA) specifications for biochemical oxygen demand (BOD). These incubators cover a temperature range ideal for BOD testing and other low temperature applications. Excellent uniformity allows for effective space utilization, assuring all areas are maintained at proper temperatures.
Thermo Scientific RT2 Basic Stirring Hot Plate is an economicalanalog unitideal for routine applications. The hot plate is designed to save bench top space. Offers overheat prevention circuit which turns off the heater if the temperature reaches 842°F (450°C), preventing accidental damage to the unit. Thermo Scientific RT2 Basic Digital Hot Plate
Thermo Scientific Compact Digital Rocking Shaker. Ensure gentle sample agitation with this compact, adjustable-angle unit. The shaker is ideally sized for placing in incubators, chambers, and refrigerators (up to 60°C). PID control ensures smooth motion.
View all of our new products.
Be the first to comment on "Take Three: Timely New Technologies from Thermo Scientific"