Our Technical Support team gets thousands of questions a year about product selection, compliance, troubleshooting issues—and just about anything a customer can think up.
Here are a few of the answers we have for some of our common products:
VP Handheld Mini Ultraviolet UV Lamps
Q: I’m looking for a UV lamp that can help me tell non-organic and organic materials apart (in glue). Would this be a good choice?
A: A longwave 365 nm UV light lamp will aid in the detection of organic materials.
Q: Can it be used for fraud detection to check the authenticity of documents, passports?
A: A 365 nm wavelength is used for fraud detection of credit cards, documentation, and older currency. This lamp would emit at the appropriate wavelength, and would work for this specific operation.
Wireless Dual Display Rain Gauges
Q: Does this rain gauge measure rainfall intensity?
A: The gauge will not measure intensity, and will only measure total rainfall. The meter can read a current rainfall total, but time will also need to be noted supplemental to the device.
Do you have a question? Call our technical support team at 1-800-323-4340.
Be the first to comment on "UV Lamps and Rain Gauges—Technical Support Answer Your Questions!"