Why Use a Thermohygrometer?
Thermohygrometers are the most common type of humidity sensor and you’ll find them in handheld units and transmitters. They commonly display temperature and humidity simultaneously and have a response rate of about four seconds. Their sensors read resistance or capacitance. Typical applications include:
Extech RH390 Precision Thermohygrometer
Extech RH390 Precision Thermo-hygrometer
  • Breweries and fermentation
  • Environmental research
  • Food ripening and storage
  • HVAC
  • Pharmaceutical incubators
  • Pharmaceutical tablet coating
  • Plant growth chambers
With features that can include large, back-lit displays, calculation of dew point, min/max, data hold, data logging, infrared temperature measurements, RS232 output, printing capabilities, and more, thermohygrometers can be handy instruments. Why focus your search on a thermohygrometer? Here is an overview of the pluses and minuses of these instruments:
Advantages to Using a Thermohygrometer
  • Low cost
  • Quick sensor response time
  • Repeatability at ambient RH values
  • Portability
Drawbacks to Using a Thermohygrometer
  • Susceptibility to losing calibration
  • Level of accuracy is medium or moderate
  • Probes are not interchangeable
View all thermohygrometers
Be the first to comment on "Why Use a Thermohygrometer?"