Clean Wipes Selection Guide
clean wipes
How to find the best clean wipe for your needs.
Let’s face it. Cleaning isn’t the most exciting task, but a necessity for every lab and cleanroom facility. Cleaning products keep equipment running at peak efficiency, reduce the risk of contamination, and ensure the health and safety of personnel. To get the job done faster and most efficiently, you can use cleaning wipes and cleanroom wipes. They are ready to use and reduce struggling with solutions and separate towels. Here are a few options that are in stock and ready to ship along with a link to our wipes selection guide, so you can find the right wipe for your application.
Critical task wipes
Cole-Parmer Essentials Critical Task Wipes are eco-friendly lab wipes made from 100% recycled cellulose. Ideal for optics and other sensitive surfaces. Perfect wipe for light-duty tasks and delicate surfaces. They will not scratch, and can be used in a variety of industries. The wipes are made of single-ply, absorbent, and soft material making them ideal for cleaning small parts, lenses, and other equipment in your lab facility. Since they are low linting and low in chemical extractables, they’ll leave surfaces extremely clean.
clean wipes
Pre-wetted clean wipes
Cole-Parmer Essentials Pre-Wetted Clean Wipes are available either with a blend of pure, reagent-grade 70% isopropyl alcohol and 30% reagent-grade deionized water, or 100% pure reagent-grade deionized water. Individually dispensed wipes are handy and ready for instant use. Snap-on sealed top eliminates contamination from dust or dirt. Each canister is provided in shrink-wrap or a sealed bag to maintain cleanliness for use in clean rooms. Wipes are 6 x 7.5 inches and packaged 100 per polyethylene canister.
Cleaning products
When wipes aren’t enough to get the job done and you need extra help, additional cleaning products we have a variety of cleaning solutions for general and critical cleaning procedures including spray cleaners for disinfecting large hard surfaces; cleaning solutions for surface cleaning, glassware, and equipment cleaning; and hand cleaners and sanitizers to help prevent diseases. Some of our cleaners are appropriate for applications that require rigorous cleaning protocols due to industry requirements. You can also find mops, brushed, waste disposal and more.
Clean wipes selection guide
Need help selecting a wipe for your needs? Use our Clean Wipes Selection Guide where we have compiled features of various types of wipes as well as their performance for specific application areas.
Related Articles
4 Types of Lab Equipment to Replace when Spring Cleaning
Laboratory Supplies to Help You Spring Clean Year Round
Get the lastest blog posts delivered to your email - sign up today
Be the first to comment on "Clean Wipes Selection Guide"