Centrifuges are instruments that use centrifugal force to separate colloidal particles or substances of different densities, to remove moisture, for microfiltration, or to simulate gravitational effects.
Centrifugation is one of the most important and widely applied research techniques in biochemistry, cellular, and molecular biology. Current research applications rely on isolation of cells, subcellular organelles, and macromolecules, often in high yields.
Features to Consider
From personal microcentrifuges to general purpose and specialty centrifuges, you already have a choice to make in terms of centrifuge types. Here are a few features you may want to consider as well:
- Anticorrosive material: Carbon fiber is durable and lightweight, and will hold up longer over time than metal or aluminum components.
- Compact footprint: A compact centrifuge can save you valuable bench space.
- Refrigeration: For samples requiring consistent temperatures, your centrifuge should be able to run at its maximum speed and still maintain a consistent temperature.
- Rotor versatility: If you perform multiple tasks, choose a centrifuge that accommodates a variety of adapters. Select a model that doesn’t require the use of tools and that is ergonomically sound.
- Throughput: Look for versatility in rotor and adapter options so that you can use your centrifuge to its optimal capacity.
Be the first to comment on "Centrifuges In the Laboratory"