Lab Safety Supply Instruments to Protect Against Hazards in the Workplace
Does your workplace have the lab safety supply instruments it needs to ensure worker safety and production efficiency? Here is a checklist of items:
Air Sampling Pumps for collecting gas samples for analysis or evaluating employee particulate exposure levels.
Combustion Analyzers measure and tune furnace/boiler efficiency to reduce fuel costs. Emissions analyzers also detect potentially hazardous gas emissions.
Fixed Gas Monitors use single-point, fixed systems to monitor for a particular gas. Gas options include oxygen, carbon dioxide, chlorine, ozone, and more.
GasAlertQuattro 4-Gas Detector
GasAlertQuattro 4-Gas Detector
Gas Detectors from detector tubes or personal, single-gas, multi-gas, and confined space monitors.
Indoor Air Quality Meters from standard models measuring carbon dioxide and temperature to fully functional IAQ meters designed for industrial hygiene studies.
Light Meters verify lighting levels are appropriate for parking lots, employee workspace, and emergency egress. Single-scale, dual-scale, and data logging models are available in a variety of meter styles.
Leak Detectors are ideal for pinpointing small leaks in boilers, furnaces, refrigeration and other systems that can emit dangerous gases.
Radiation Meters to address concerns about alpha, beta, or gamma radiation in your laboratory. Detect surface or area radiation levels.
Sound Meters/Dosimeters measure dB noise level in work or production areas to ensure levels do not exceed OSHA standards. Dosimeters also measure individual worker exposure over time.
Also consider:
Matting to prevent worker fatigue, slips, and falls in a wide variety of mat materials and styles for laboratory, food, pharmaceutical, chemical, or industrial applications.
Right-to-Know and Signs keep workers informed of workplace hazards. See a variety of MSDS centers, posters, labels, signs and more.
Lockout / Tagout products to keep your company in compliance with OSHA regulations. Choose from a wide variety of tags, hasps, complete kits, and more.
1 Comment on "Lab Safety Supply Instruments to Protect Against Hazards in the Workplace"
  1. Good information here. I noticed you mention OSHA regulations, if a company is doing international work, I’d suggesting getting ISO certified as well.