Chemical Hose that Meets the Challenge
Terminator Multi-Purpose Hose
With broad chemical resistance, chemical hose is used to transfer chemical compounds. With many types to choose from, here is an overview of some common hose options and the applications to use them for:
  • General purpose hose when transferring oil is not a significant factor
  • Multipurpose hose for petroleum oils, gasoline, kerosene, fuel oil, lubricating oils, air, and water
  • PVC and Industrial PVC hose for basic vacuum or transfer and high flow rate applications
  • Smooth PTFE-lined PTFE hose for sanitary or steam transfer, bottle filling, gas analysis, hydraulic lines, extrusion presses, molding and adhesives
  • Smooth FEP-lined PTFE or rubber hose for bulk transfer, chemical trailer loading, portable pumps, isolation dampeners, food & beverage, flavors, load cell applicators, caustic cleaning, CIP
  • Convoluted FEP-lined hose for chemical, steam, solvent, food & beverage, acid lines, CIP lines
  • Rubber-covered Chemfluor® hose for pharmaceutical, chemical, food and beverage handling
  • Platinum-cured SS wire-reinforced silicone hose for demanding sanitary high pressure and high vacuum, pharmaceutical, food and beverage, and cosmetic applications
  • Polypropylene-braided convoluted Chemfluor PTFE hose for chemical and industrial uses; tightly woven braid reinforcement resists abrasion better than stainless steel reinforcements
Be the first to comment on "Chemical Hose that Meets the Challenge"