There’s more to collecting environmental samples than it seems. From the type of container to what should (or should not) be added to how to store it—all can determine if what you analyze is an accurate measure of what’s in the sample.
Once the sample bottle is clean enough for use, you must select the proper material to use. Different parameters have different requirements.
Most organic parameters must be collected in glass while inorganic parameters can use plastics or fluoropolymers as well. Because not all plastics have the same properties or inherent background levels, you are typically limited to polypropylene, LDPE, or polyethylene. Bacteria samples are generally limited to materials that can be sterilized. Also, you may have requirements concerning fluorescence or clarity depending on the analytical method you use.
Using the most efficient laboratory supplies can make your process easier. Here are products to assist in sample collection, preparation, and storage:
- Environmental Bottles: a full selection of certified, pre-cleaned bottles, glass bottles, cylindrical options, and more
- Disposable BOD Bottles: Never wash a bottle again—these are lightweight, precleaned, and EPA-approved
- ProWeigh® Filters: No prep needed—get improved accuracy and quicker reporting
- FlipMate® Filtration Systems: Fast and easy—filter samples in second
- Digestion Cups: For HotBlock® metals digestion—graduated to dilute, digest, and store in one vessel
View our selection of efficiency-enhancing products for the environmental lab.
Be the first to comment on "5 Ways to Enhance the Environmental Sample Process"