Tabletop Filling systems are ideal for the repetitive dispensing of free-flowing, semi-viscous (such as syrup, lotion, or thin oils), or viscous fluids (such as paste, cream or detergent). They can dispense solutions for pharmaceutical small batch filling needs with precise accuracy and a high cycle rate.
The liquid is drawn in by the intake stroke and dispensed by the discharge stroke. Volume control enables you to increase or decrease the desired dispensing volume. The system operates via a footswitch or continuous mode cycle.
Single-head or dual-head filling stations are available in tabletop models. Models may vary in their power capacity—choose the VAC that suits your application.
\
Be the first to comment on "Spotlight on Pumps: Filling Pumps"