To maintain a constant temperature of from -50°C to 280°C in your bath, use bath fluid to match your application. Select from the following options:
| Bath Fluid | Temperature Range | Specific Gravity |
| Dynalene® HC-50 fluid | -50 to 58°C | 0.85 |
| Silicone 210 fluid | -65 and 280°C | 0.98 |
| Silicone 200 20 (hot) bath fluid | 10 and 149°C | 0.95 |
| Silicone 710 (hot) bath fluid | 150 and 250°C | 1.11 |
To reduce significant vapor loss and provide insulation, use hollow ball bath covers. These ball covers are made of HDPE and form a tight cover over your fluid. Hollow bath balls can reduce heat exchange by more than 75%.
To hold your sample in place, check out holders and tube racks which protect bath fluid.
See our full line of bath and circulator accessories.
Be the first to comment on "Controlling Bath Fluid Temperatures"