Does your drinking water taste funky?
Use a drinking water test kit.
Fast, easy-to-use water test kits detect hazardous contaminants in municipal and private well drinking water.
  • Lab-quality results in a simple-to-use, affordable test kits
  • Compare your results with EPA-recommended levels
  • Comprehensive test kits detect most common water contaminants
Drinking water test kits indicate the presence of bacteria, pesticides, and lead. Kits also include readings for nitrate (0 to 50 ppm), nitrite (0 to 10 ppm), total chlorine (0 to 10 ppm), pH (6 to 10), and water hardness (0 to 425 ppm). Well water test kits add tests for contaminants commonly found in private well water: iron and copper (0 to 5 ppm). Order bacteria, pesticides, and lead stand-alone kits if less comprehensive tests are required. All kits are for one-time use. Don’t wait until someone gets sick. Buy one today.
Be the first to comment on "Does your drinking water taste funky?"