With July as National Ice Cream Month, we turn our attention to some savvy equipment that can help prepare delicious treats. Specifically, today’s focus is mixers. Cole-Parmer offers a broad selection of mixers and mixer products for various applications. Selecting an appropriate mixer is easy once you consider the variables:
- Container capacity: tank diameter and batch height
- Liquid viscosity: different types of liquids display different characteristics when force is applied. Four most common types of liquid behavior are listed below—all of our mixers are to be used with fluids exhibiting Newtonian, pseudoplastic, or thixotropic behaviors.
- –Dilatant Liquids—viscosity increases as shear rate increases. Mixers can bog down and stall after initially mixing such liquids. Dilatant liquids include slurries, clay, and candy compounds. View Real-Torque Digital Brushless Mixers.
–Newtonian Liquids—viscosity remains constant regardless of shear rate or agitation. As mixer speed increases, flow increases proportionately. Newtonian liquids include water, mineral oils, and hydrocarbons. See the Cole-Parmer® Stir-Pak® General Purpose Laboratory Mixers.
–Pseudoplastic Liquids—viscosity decreases as shear rate increases, but initial viscosity may be sufficiently great to prevent mixing. Typical pseudoplastic liquids are gels, latex paints, and lotions.
–Thixotropic Liquids—as with pseudoplastic liquids, viscosity decreases as shear rate or agitation increases. When agitation is stopped or reduced, hysteresis occurs and viscosity increases. Often the viscosity will not return to its initial value. Thixotropic liquids include soaps, tars, shortening, glue, inks, and peanut butter. - Torque requirements: the rotational force required of the mixer motor—measured in in-oz or in-lb. Torque and speed are inversely related (as speed increases, torque decreases).
- Horsepower (hp) requirements: the efficiency required of the mixer motor with regard to torque (in-oz) and to the rotation speed (in revolutions per minute—rpm) as defined by the following equation: hp = (in-oz x rpm)/1,008,400
- Rotational speed (rpm) and diameter of mixing propeller: A small increase in the rpm or diameter greatly increases the power required for mixing. This relationship can be expressed as follows: Power α rpm3 x dia5
- Duty cycle: the time interval devoted to starting, running, stopping, and idling when a device is used intermittently.
- Power supply: electric (115 or 230 VAC) or air-drive mixers for areas where electricity is unsafe or impractical.
Be the first to comment on "For Ice Cream and Other Treats, Choose the Right Mixer"