When you need to get hard metals, glass, plastic, and ceramics really clean, an industrial ultrasonic cleaner is your go-to device. It uses sounds waves between 20 and 400 kHz to clean, along with a solvent specific to your application. Get greater cleaning action from solvents with lower surface tensions. Use a solvent—perhaps an organic solvent—that can dissolve your contaminants.
How does it work?
An industrial ultrasonic cleaner works fast—processing time is typically about two to six minutes. The cavitations caused by the ultrasonic waves do the cleaning. Note that:
- A higher frequency industrial ultrasonic cleaner generally cleans in greater detail and a bit more gently
- A low frequency model is usually considered a coarse instrument
Automotive shops, research labs, printers, and semiconductor manufacturers all use ultrasonic cleaners.
View our selection of ultrasonic cleaners.
Be the first to comment on "Get Uniform Cleaning with Ultrasonic Cleaners"