Life gets noisy and the products around us add to that noise. Reducing sound levels begins with testing products with sound meters to assess how much noise they are emitting. Sound meters also assess how much environmental noise we are exposed to—and whether it exceeds compliance levels.
Get precision sound decibel measurements for product noise certification and reduction applications by using a data logging sound level meter. By comparing differently weighted measurements, you can isolate the source of problem sounds. Units featuring A and C weighting capabilities measure noise levels ranging from industrial machinery or equipment to residential and commercial environments.
To analyze data, study it for trends, and then generate reports, you’ll want a meter that includes software. Some advanced units also have a timer function to control the start and end of a run for automatic recording as well as event-based threshold noise level to trigger recordings above a significant level.
View our selection of sound meters, including kits, dataloggers, and more.
Be the first to comment on "Measure Noise with Sound Level Meters"