Hot Product Alert! Homebrewers love the Love temperature controller
Homebrewers
Homebrewers love this Love Temperature Controller. These controllers are ideal for on/off control applications where a digital display and an adjustable deadband are needed over a limited temperature range. Use with packaging equipment, food cooling and chilling equipment, wine cellars, and more!
Each controller has a tactile front keypad that lets you program 12 different parameters, including set point, deadband, cycle time and ambient probe adjustment. Programmable password protection prevents accidental changes to settings. The bright red, 3-digit LED display is easy to read even in dim areas.
Read our community contributed product reviews and Questions and Answers.
View all of the details on this temperature controller.
Be the first to comment on "Hot Product Alert! Homebrewers love the Love temperature controller"