Hot product alert! Use our Cole-Parmer SpatulaBalance to scoop and weigh instantly.
Cole-Parmer® Traceable® SpatulaBalance™
Check out this hot product alert!  This product combines a scoop and a balance to speed up your time in the lab and eliminate spills.
Cole-Parmer® Traceable® SpatulaBalance™
  • Unique spatula both scoops material and instantly displays weight
  • Scoop design eliminates spilling
  • Three-position soft-touch keys in the handle tares (sets to zero), holds (freezes the display), and changes grams/ounces
BUY ME on coleparmer.com.
Cole-Parmer SpatulaBalance™, 300 g x 0.1 g 
Be the first to comment on "Hot product alert! Use our Cole-Parmer SpatulaBalance to scoop and weigh instantly."