Lab Equipment: Water Purification Systems and Methods of Purification
Thermo Scientific Barnstead Pacific RO Water Purification System
Thermo Scientific Barnstead Pacific RO Water Purification System
Water purification lab equipment ensures the quality of the water used in the lab to perform successful and reliable tests. Many water purification systems can handle a range of applications from basic buffer preparations to highly sensitive protocols such as HPLC, PCR, or uses that require precise TOC removal and measurement. Other systems can be used for rinsing lab glassware, supplying autoclaves and ultrapure water systems, and preparing and diluting buffers, reagents, and media.
Water grade standards are classified as Type 1 (ultrapure water, 18.2 MΩ), Type II (pure water, 1 to 15MΩ), Type III (reverse osmosis water) and Type IV (cartridge/filter system water). Water feed sources can range from tap water to distilled, deionized, or reverse osmosis (RO) water).
Methods of Purification:
Adsorption: Activated carbon bonds with the chlorine and organic materials in feedwater to immobilize and remove these impurities
Deionization: The removal of ions and minerals by synthetic ion exchange resins. Cation resins remove positively charged ions; anion resins remove negatively charged ions
Distillation: The process in which water is heated to a gaseous state and recondensed in a separate vessel
Filtration: Use as a pretreatment or a stand-alone treatment. Water passes through a filter of specified porosity at normal line pressures. The filter retains most particulates, with water passing through
Reverse osmosis (RO): Use primarily as a pretreatment. Equal amounts of pure water and saline solution are separated in a U-tube by a semipermeable membrane. When external pressure is applied to the saline side, the semipermeable membrane allows water to pass through while salts are concentrated and flushed down a drain
Ultrafiltration: Use for removing pyrogens and bacteria. Under pressure, water is forced through a membrane with a pore size smaller than 0.005 μm. Particulates are retained, with only pure water passing through
Ultraviolet (UV) oxidation: UV light (at <280 nm) passes through the water destroying bacteria, viruses, and trace organics
See our selection of Water Purification Systems.
7 Comments on "Lab Equipment: Water Purification Systems and Methods of Purification"
  1. Thanks for sharing really informative article about water purification and its process. I was looking for such kind of article. Keep sharing such informative article.
  2. Dear Sir,
    Kindly send us your quotation for supplying the below required Lab. equipment for water treatment systems.
    The required lab. equipment are segregated in two requirements
    1- Critical Lab. equipment within each water treatment system.
    2- Water Quality Lab. equipment.
    The equipment selection must be economical and there is no problem if multi parameter device is quoted.
    if you are interested, pls confirm back to send you the details.
    Best Regards
  3. May you please send me a quotation for the Reverse osmosis equipment to use in the Lab to purify water for use during analysis
  4. Dear Sir,
    My name is Ahmad Smadi the proposal manager at Miahona, Miahona is a developer, owner and operator of municipal water and wastewater based in Saudi Arabia.
    We are looking for Laboratory MBR package 1 or 2 m3/day.
    Regards
    Ahmad Smadi
  5. su arıtma cihazı | March 29, 2023 at 1:59 am | Reply
    thanks for share good post