Laboratory Equipment: A Guide to Mixer Selection
When choosing laboratory equipment such as a mixer, several variables should be considered specific to your application. For mixers, gather the follow information:
Stir-Pak High-speed, Low-Torque Mixer System (23-2300rpm, 115 VAC)
Stir-Pak High-speed, Low-Torque Mixer System (23-2300rpm, 115 VAC)
  • What container capacity do you need?
  • What liquid viscosity?
  • What are your torque requirements?
  • What are your horsepower requirements?
  • What rotational speed or RPM do you need? What diameter for the mixing propeller?
  • What are your requirements for duty cycle?
  • What will be the source of your power supply?
Laboratory Mixers are available with permanent magnet, universal, or air-drive motors. Choose different mixer mounts for mounting on tanks or stands. A variety of impellers and paddles lend versatility to the mixer you select.
Industrial Mixers are available with electric- or air-driven motors for up to 1-hp. Fixed- or variable-speed and direct- or gear-driven options are available.
Specialty Mixers include:
Sanitary Mixers for all applications requiring extra clean processes.
Bulk-Container Mixers mount atop tote containers and mix liquids that are in long- or short-term storage.
Handheld Mixers for mixing thin or heavy liquids in open or closed containers.
Static Mixers are installed inline and mix fluids as you pump them through a pipeline. The fluid moves through the mixer in an alternating clockwise/counterclockwise motion to ensure a homogeneous product. They are virtually maintenance free and need no spare parts.
Our Application Specialists can assist you in choosing the right impellers for optimum mixing. View our mixer selection.
Be the first to comment on "Laboratory Equipment: A Guide to Mixer Selection"