Laboratory Ovens: What’s the Difference?
StableTemp® Vacuum Oven
StableTemp® Vacuum Oven
If you are drying, softening, ageing, or sterilizing, you have a choice of ovens and heating technologies to do the job. Here’s an overview of the various types:
Gravity Convection Laboratory Oven:
StableTemp® Vacuum Oven
StableTemp® Vacuum Oven
  • Low cost alternative (to mechanical convection ovens) when temperature uniformity is not paramount
  • Use when a powder or substance to be heated or dried would be disturbed by a fan-generated airflow
Mechanical Convection Laboratory Oven:
  • Incorporates a motorized fan to push air across the heating elements and into the chamber
  • Fan enables more consistent distribution of air
  • Designed for good temperature uniformity or faster heatup/drying time
Safety Oven:
  • These mechanical convection ovens contain a blow-out panel to protect you from injury if an explosion occurs within the oven
  • NOT explosion-proof —will not prevent volatiles from combusting
  • If an explosion occurs, the blow-out panel will direct the force of the blast away from you
Vacuum Oven:
  • Use when you need a lack of atmosphere or inert atmosphere
  • Heating occurs in a vacuum where there is no convection of air. Place the sample of the shelving in the unit so heat is conducted from the side walls to the sample via the metal shelf
  • Requires a separate vacuum pump
View our full selection of ovens.
Be the first to comment on "Laboratory Ovens: What’s the Difference?"