Using Two Using Two Mixers for High-Torque, Large-Batch Mixing? Switch to One: Watch Video
Cole-Parmer Digital Batch Mixer
Cole-Parmer Digital Batch Mixer (Item #50009-00)
Mixing viscous fluids in large batches can be a challenge. In fact, you might be using two mixers to get the job done. Here’s a product that eliminates the need for two mixers, saving you time in the lab. The Cole-Parmer digital batch mixer has enough power to mix up to 52 gallons (200 L)! It offers one of the highest torque settings on the market today—4248 in-oz (30000 N-cm).
Ideal for laboratory and industrial uses, the mixer features RS-232 and USB interfaces to record date, time, speed, and torque data. Easy-to-use resistive digital touch-screen control enables you to select time and speed with a gloved hand or pencil tip. Other functions include alarms, timers, torque zero, torque min/max setting, and clockwise and counterclockwise rotation.
Watch the video below for more information on the latest digital batch mixer from Cole-Parmer.
Shop our entire line of mixers and blenders.
Be the first to comment on "Using Two Using Two Mixers for High-Torque, Large-Batch Mixing? Switch to One: Watch Video"