The Jenway 72 scanning spectrophotometer series leverages diode array technology to produce exceptionally fast results. Simultaneous measurement of all wavelengths increases productivity with a scan time as short as 3 seconds. Now that will save you time in the lab! These spectrophotometers are ideal for applications in education and routine testing in clinical, veterinary, pharmaceutical and QC laboratories.
Watch how it performs a spectrum scan.
Watch the Jenway 72 Series Simple Photometric Measurement video.
Watch the Jenway 72 Series How to Save Results video.
Browse our complete line of spectroscopy and spectrophotometers.
Be the first to comment on "Got a Minute? Watch a Spectrophotometer Perform a Spectrum Scan in 3 Seconds"