What’s smart, interchangeable, and tests multiparameters? It’s the new Smart Meter Technology with digital “Intelligent Design” sensors in the YSI® MultiLab DO/BOD Benchtop Meter Kits . The “plug-and-play” feature enables you to switch electrodes to test for pH/ORP, conductivity, and optical DO/BOD as well as operate the unit as a conductivity meter.
For water and wastewater applications and quality control for the food & beverage, pharmaceutical, and chemicals industries, the Smart Meter Technology offers several advantages.
It complies with US EPA regulations by meeting approved methods for dissolved oxygen and BOD. With highly accurate measurements, it displays readings in large graphics on an easy-on-the-eyes backlit screen. Manual and automatic data logging has GLP traceability. The CMC function continually monitors the sensors, measuring range against calibration range. The sensors digitally store their serial number, unique ID, and calibration data and transfer this data when moved.
View the YSI® MultiLab DO/BOD Benchtop Meter Kits, available in single-channel meter, two-channel meter, or three-channel meter formats.
Browse All Dissolved Oxygen Meters available at Cole-Parmer
Be the first to comment on "Using New Technology to Switch Electrodes to Test Water"