What’s in Your Water? Types of Water Testing Equipment
Oakton product group
Water testing is the continuous sampling of various liquid streams and the analysis of their quality. These liquid streams include watercourses such as rivers and lakes, groundwater, recirculated cooling streams, boiler feedwater or condensate, and process effluents. Water testing is not only a vital element of preserving a safe public drinking supply and a healthy environment, it can also improve industrial process efficiency for better products at reduced cost. Our selection of water testing equipment includes test kits for bacteria and chemical compounds; colorimeters; multiparameter meters for measuring pH, ORP, conductivity/salinity/total dissolved solids (TDS), dissolved oxygen (DO), and temperature; and turbidimeters.
Bacterial Test Kits
Eliminate time-consuming culture/plating techniques for convenient bacterial counts and assays.
Colorimeters
Measure the light absorbed by your sample after it reacts with a reagent that produces a color change. Since the intensity of absorbed light is proportional to the concentration of the compound, colorimeters accurately indicate the concentration of various compounds in your sample. Many colorimeters work with specific test kits for easy analysis. This reduces operator error, whether in reading color changes or in measuring reagents.
Multiparameter Meters
Measure pH, ORP, conductivity/salinity/total dissolved solids (TDS), and/or dissolved oxygen (DO)/biological oxygen demand (BOD).
Test Kits for Chemical Compounds
Contain prepared reagents and standards that let you easily determine the concentration of specific compounds in the sample. We offer titrimetric and colorimetric test kits for several compounds including ammonia, chlorine, carbon dioxide, heavy metals, and many others.
Colorimetric test kits:
 These tests determine the concentration of a substance. The higher the concentration of a substance, the darker the color developed in the test.
Other elements can interfere with the accuracy of the tests. Read kit instructions for appropriate adjustments.
Titrimetric test kits:
The concentration of a substance in a sample solution can be determined by titrimetric tests. After the sample is treated with an indicator, a standard titrant is added until a color change indicates a completed reaction. The amount of titrant used corresponds to the concentration of the substance being tested.
Turbidimeters
Turbidity in water is caused by suspended and colloidal matter such as clay, silt, finely divided organic and inorganic matter, and plankton and other microscopic organisms.
Quantify the turbidity of your sample by measuring the light scattered at a 90¡ angle to a source light beam. The meter compares the light scattered by sample particulates to light scattered in a fluid with known turbidity and displays the measurement in nephelometric turbidity units (NTU). The presence of dissolved, color-causing substances that absorb light may cause a negative interference.
Turbidimeters with scattered light detectors located at 90° to the incident beam are called nephelometers. Nephelometers are relatively unaffected by small differences in design parameters and therefore are specified as the standard instrument for measurement of low turbidities. Poor measurement technique can have a greater effect on measurement error than small differences in instrument design.
Determine turbidity as soon as possible after the sample is collected to ensure examination under original conditions.
  1. Gently agitate samples to ensure a representative measurement. If storage is required, cool to 4°C to minimize microbiological decomposition of solids.
  2. Keep cells scrupulously clean, inside and out, and discard if scratched or etched. Never handle them near the instrument’s light beam.
  3. Fill cells with samples and standards—allow sufficient time for bubbles to escape. Because small differences between sample cells can significantly impact measurement, use either matched pairs of cells or the same cell for both standardization and sample measurement.
Coagulation-Flocculation Jar Test of Water: This is a general procedure for the evaluation of a treatment to reduce dissolved, suspended, colloidal, and nonsettleable matter from water by chemical coagulation-flocculation, followed by gravity settling. The practice provides a systematic evaluation of the variables normally encountered in the coagulation-flocculation process. The coagulation-flocculation test is carried out to determine the chemical, dosages, and conditions required to achieve optimum results. The primary variables to be investigated using the recommended practice include, (but are not limited to): chemical additives, pH, temperature, and the order of addition and mixing conditions.
Significance: This jar test permits the evaluation of various coagulants and coagulant aids used in the treatment of water and wastewater for the same water and the same experimental conditions. The effects of the concentration of the coagulants and coagulant aids and their order of addition can also be evaluated by this practice.
Related Articles:
Good Laboratory Practices for Taking Turbidity Measurements
Water Purification
28 Comments on "What’s in Your Water? Types of Water Testing Equipment"
  1. Hi: I am interesting in water examination. I need simple operative equipment for water analysis
    water testing equipment includes test kits for bacteria and chemical compounds; colorimeters; multiparameter meters for measuring pH, ORP, conductivity/salinity/total dissolved solids (TDS), dissolved oxygen (DO), and temperature; and turbidimeters. How much they cost in dollars. please send me the prices. Thank you.
  2. water color change after 3 to 5 hrs as yellow at my home water pump
  3. Hi, we would like to know the equipments or a test kit used for the microbial testing of the water quality.
    • Hello:
      I forwarded your question to our team in India to help you with your question. If you need immediate assistance, you can call 022-6139-4444. Thank you.
  4. Hi, I would like to know what are the equipments are used in water testing laboratories
  5. I am impressed by the range of equipment and kits you have. Do you have an agent in Kenya?
  6. Hi, we have requirement of water quality monitoring device in India, could you let your team here get in touch with me for sharing further details.
    • Cole-Parmer Blog Team | October 8, 2020 at 6:32 am | Reply
      Hello Abhi:
      I sent your info to our India expert and someone will definitely be getting back to you. What is the best way for someone to get in touch with you?
  7. We have a hot artisan spring approximately 2.46 cfs flow. I would like to test for bacteria and fecal matter, hard minerals, ph, suspended particulates in our spring. Wondering about type and cost of equipment
    Thank yiy
  8. Send to me necessary water laboratory equipment . We have an intention of setting up water laboratory for public water supply and irrigation in our University in Nigeria. Thanks you.
    • Antylia Scientific Blog Team | April 7, 2021 at 5:08 pm | Reply
      Hi Prof. Oluyege:
      I sent your request to our technical experts. They will reach out to you. In the meantime, should you need immediate assistance, please call us at 1-800-323-4340. Thank you.
      K. Danko
  9. amrou mohammed | January 5, 2022 at 1:59 am | Reply
    Good evening
    We are looking for a public water quality and safety meter
    Instant and portable site measurement
    looking forward to measuring
    pH
    turbidity
    Total dissolved salts
    coliform bacteria
    ammonia
    • Antylia Scientific Blog Team | January 5, 2022 at 1:35 pm | Reply
      Hello:
      Thank you for your interest. I forwarded your request to our technical expert. Someone will be emailing you. Thanks!
      Karen Danko
  10. edgar I. peque | January 9, 2022 at 11:35 pm | Reply
    Hi, I would like to know what are the available equipments for water physical quality testing like pH, ORP, conductivity/salinity/total dissolved solids (TDS), and/or dissolved oxygen (DO)/biological oxygen demand (BOD).
  11. Interested in testing for bacteria and would like a full product guide please.
  12. Ferdinand Villoso | April 4, 2022 at 11:07 pm | Reply
    Im interested in putting up a water testing laboratory that would cater water bacteriological examinations and physico-chemical test, i would you send me a qoutation of equipment and machines needed for such set up. Thank you very much
  13. Hello ! thanks for the brief explanation. I am making a list of laboratory equipment for water quality testing like, bacteria kit, colorimeter, multiparameter meters, test kits for chemical compound, colorimetric test kit, turbidimeter. Can you help me pick up all necessary equipment used for water quality testing, thanks !
    • Antylia Scientific Blog Team | October 18, 2022 at 11:12 am | Reply
      Hello Melaku:
      Thanks for reaching out to us for help with this task. We will have one of our technical experts work with you to gather your list.
      Karen
  14. Victoria Venuso | January 21, 2023 at 11:12 pm | Reply
    hello,
    I am currently in search of commercial water testing equipment that we can use in the Philippines that can do microbiological analysis and Physical and chemical analysis for drinking water
    • Antylia Scientific Blog Team | January 23, 2023 at 11:37 am | Reply
      Hi Victoria:
      Thanks you for reaching out to us. I will forward your request to our technical experts and someone will contact you with more information. Thanks!
      Karen