Why Use a Recirculating Chiller?
Cole-Parmer® Polystat® Recirculator with Force/Suction Pump
Cole-Parmer® Polystat® Recirculator with Force/Suction Pump
Among baths, circulators, and recirculating chillers, why choose the latter? <a href=”/c/recirculators-and-chillers>Recirculating chillers deliver a flow of temperature-controlled fluid. They are enclosed circulators featuring robust cooling systems used for demanding continuous-use lab applications.
Cole-Parmer® Polystat® Recirculator with Force/Suction Pump
Cole-Parmer® Polystat® Recirculator with Force/Suction Pump
Recirculating chillers offer:
  • Temperature control from -10° to +80°C
  • Sealable reservoirs which permit open-loop and closed-loop use
  • Pumping capacities up to 15.5 L/min
In addition to many lab applications such as chemical, pharmaceutical, and food and beverage processing, chillers are used in industrial settings. These include:
Plastics: chillers cool the hot plastic that is injected, blown, extruded, or stamped. Chillers can also be used to cool down the equipment used in the manufacturing process.
Lasers: chillers cool down lasers and the power supplies used to power them.
Printing: chillers remove the heat generated by the printing rollers and also cool down the paper after it comes out of the ink drying ovens.
Machine tooling: chillers cool the spindle of the machine as it produces the part and cools the liquid being sprayed on part itself as it is being turned on the spindle.
View our <a href=”/c/recirculators-and-chillers>selection of compact chillers, flow-through refrigerated chillers, refrigerated chillers with immersion probe, immersion coolers, temperature-controlled recirculators, and recirculating chillers.
2 Comments on "Why Use a Recirculating Chiller?"
  1. Good Morning Cole-Parmer Team,
    To introduce myself, I am Harsh Shah, a PhD student at Long Island University Brooklyn, NY.
    We are looking for a price quote of recirculating chiller for Hot Melt extruder with below mentioned specifications –
    1. Reservoir capacity – 24- 30 Lit
    2. Cooling Capacity – 2.5 kW (@ 20 C)
    3. Fitting size : 3/4″ ID (Barbed) and pump connectors – G 3/4″. (This are requested considering the broader diameter which can be reduced later or changed)
    4. Temp range – -20 C to +40 C
    5. Pump flow rate – 40 L/min
    6. Pump pressure – 7.3 – 43.5 PSI
    I shall await for your reply.
    In case of any concern, I can be reached at [email protected] or 862-368-4439
    Regards,
    Harsh Shah
    • Hi Harsh:
      Thanks for your request. You should be hearing from our Quotes team shortly.
      Kind regards,