3 New Laboratory Convection Ovens for Efficiency and Economy
Cole-Parmer® StableTemp™ Laboratory Oven
Cole-Parmer® StableTemp™ Laboratory Oven
In pharmaceutical, chemistry, biology, forensics, and environmental labs, ovens dry and process samples, remove solvents or waters, and provide heat and stress testing. A new line of Cole-Parmer® StableTemp Laboratory Ovens offers scientists several options—from economy to digital models—to enhance their processes:
  • Digital Mechanical Convection Ovens quickly dry and heat samples. Save time with fast heat-up and uniform temperature throughout the chamber for greater drying efficiency. Program and control through PC, if desired.
  • Economy Mechanical Convection Ovens deliver forced air convection with PID temperature control. Includes auto-tuning and calibration features typically found on more expensive units. Set digital timer up to 99 hours.
  • Economy Gravity Convection Ovens provide gentle air flow for sterilization and drying. Two adjustable vents control air and vapor circulation. Preset up to three settings for quick recall.
Need more info? View our selection of laboratory ovens.
3 Comments on "3 New Laboratory Convection Ovens for Efficiency and Economy"
  1. Hurrah, that’s what I was searching for, what a stuff! present here at this webpage, thanks admin of this web
    page.
  2. What’s up, I would like to subscribe for this webpage to take most recent updates,
    so where can i do it please help out.