New Oakton Dissolved Oxygen Meters Showcase New Technology
oakton waterproof meter
OaktonWaterproofDO450MeterNew Oakton® Waterproof DO 450 and pH/DO 450 Meters use optical RDO technology to reduce maintenance time and costs. Optical meters provide the convenience of no membrane replacement, no electrolyte replacement, no incoming flow required, and no hydrogen sulfide interference. The meters also offer a short response time, minimal drift, and minimal maintenance. The durable, dustproof design is IP67-rated.
In addition to this new technology, Oakton is also introducing the following meters:
Oakton® Waterproof pH 150 and CON 150 Meters are rugged, accurate, and reliable—the best value for IP67-rated waterproof meters. View measurement with temperature on large, three-line LCD. Store up to 150 measurements along with temperature via built-in memory.
Oakton® Waterproof pH 450 and CON 450 Meters are backlit, rugged meters ideal for any environment. They offer a durable, waterproof, and dustproof design with IP67 rating. View measurement with temperature and calibration data with % slope or cell constant on large, custom display.
Oakton® Waterproof pH 450/CON 450 Meter: select meter with combination pH/CON probe or separate pH and conductivity probes. Durable waterproof and dustproof design with IP67 rating.
Switch between pH, mV, conductivity, TDS, and salinity with a press of a button.
View our full selection of Oakton water quality meters.
Be the first to comment on "New Oakton Dissolved Oxygen Meters Showcase New Technology"