Three Dissolved Oxygen Technologies: Which Works Best for You? (Part 1)
polarographic technology in use
Dissolved oxygen is a critical measurement for wastewater treatment. An operator must ensure proper aeration for the aerobic bacteria in the tank to break down organic suspended solids. Three technologies for measuring dissolved oxygen are employed in the market today: Polarographic, Galvanic, and Optical.
PolarographicPolargraphicImage_DO_35415_00_left
Clark-cell, polarographic technology supplies constant electrical voltage across the anode and cathode. As oxygen is reduced, the current supplied to the cathode is increased. The amount of electricity required to reduce the oxygen at the cathode is equivalent to the dissolved oxygen present.
This technology was developed in 1956 and is familiar to most operators. The challenges of polarographic technology include necessary warm-up time, membrane replacement, and the potential of electrolyte solutions degrading.
Galvanic
Galvanic technology, developed in 1964, consists of an anode and cathode in electrolyte solution covered with an oxygen-permeable membrane. As oxygen permeates the membrane, the cathode reduces the oxygen, creating a potential that is equal to the amount of oxygen in the system.
This method offers the advantages of eliminating warm-up times, as the self-polarized probe is ready to read. However, galvanic technology still struggles with membranes and electrolyte solutions. Interference occurs with the presence of any chemical that produces an electrical charge.
Look for part II to read about optical dissolved oxygen technology.
Be the first to comment on "Three Dissolved Oxygen Technologies: Which Works Best for You? (Part 1)"