Four Components in Creating a Freeze Drying System
Freeze drying removes water or solvents from a frozen product. The process of prefreezing, primary drying, and secondary drying is all done with freeze dryers. Typically used in the pharmaceutical industry, freeze drying helps to…