These ULT Freezers Do More Than Keep Their Cool
ULT freezer
ULT freezers with integrated Traceable data logger and TraceableLIVE
These one-of-their-kind ULT freezers do more than keep their cool, they have an abundance of features to help you keep your cool too. Get peace of mind with the TEC2 series and the TSi series—both are equipped with a TraceableLIVE-compatible data logger, integrated into the body of the freezer.
Securely monitor your critical environments and get notifications to your smartphone, tablet, or PC should your power go out and more. Now, you can ensure your valuable samples are stored safely, around the clock, 24/7.
The new North Sciences/Traceable® TSi and TEC2 Series Ultra-Low Temperature (ULT) Freezers provide total sample security and peace of mind 24/7. These self-monitoring freezers are integrated with a Traceable data logger compatible with TraceableLIVE® cloud-based monitoring services. TraceableLIVE provides audible and visual alerts for any out-of-range conditions via Apple Watch®, smartphone, tablet, or PC. It’s also easy to connect to all data using the TraceableLIVE app. These ULT freezers are ideal for use in healthcare, vaccine monitoring (Vaccines for Children (VFC) compliant), critical material storage, or anywhere valuable samples need to be stored safely. See the models below:
North Sciences/Traceable® Model TEC² Ultra Low Temperature Freezers
North Sciences/Traceable® TSi Series Ultra-Low Temperature Freezers
Ideal for use in healthcare and vaccine monitoring (VFC compliant), or anywhere valuable samples need to be safely stored
Dual cooling systems for faster pull down and door opening recovery times; one can maintain temperature if the other is disabled
Available in either 120 V, 208 V or 220 V and in 15 to 30 cu ft capacities
Get it fast — manufactured in the US and shipped direct from the manufacturer
Ideal for use in healthcare and vaccine monitoring (VFC compliant), or anywhere valuable samples need to be safely stored
Variable-speed inverter drive adapts to user patterns, reducing energy and HVAC costs without compromising performance
Available in either 120 V or 220 V and in 13 to 30 cu ft capacities
Get it fast — manufactured in the US and shipped direct from the manufacturer
View all Traceable products
Learn more about TraceableLIVE
Related Articles
Connect to Your Critical Environments 24/7 with TraceableLIVE
What does traceable mean?
Be the first to comment on "These ULT Freezers Do More Than Keep Their Cool"