Fisher Chemical Isooctane, OPTIMA™ Grade, Glass Bottle, 4 L

Representative image only.

Fisher Chemical Isooctane, OPTIMA™ Grade, Glass Bottle, 4 L

Fisher Chemical | Mfr: O3014| Item: UX-88013-22

Quality and consistency your lab can count on

$612.50USD / Each

$612.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥99.0%
  • Color (APHA)—≤10
  • Pesticide Residue Analysis (as Heptachlor Epoxide)—≤10ng/L
  • Optical Absorbance at 205nm—≤1.00
  • at 225nm—≤0.10
  • at 254nm—≤0.01
  • Fluorescence Background (as Quinine Sulfate)—≤1ppb
  • Water—≤0.01%
  • Residue after Evaporation—≤1ppm
  • Refractive Index at 25°C—1.3885 to 1.3905
  • Density at 25°C—≤0.690g/mL
  • Water-soluble Titratable Acid—≤0.0003mEq/g
  • Sulfur Compounds (as S)—≤0.005%
  • Packaged under Nitrogen, Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmfulStorage Code Red
Specifications
  • Purity GradeOPTIMA
  • Empirical Formula(CH3)3CCH2CH(CH3)2
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsOctane, Iso-, 2,2,4-Trimethylpentane
  • Volume (mL)4000
  • Formula Weight114.23
  • PackagingGlass Bottle
  • DOT InformationOctanes; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable, Harmful
  • UN NumberUN1262
  • CAS Number540-84-1
  • Boiling Point C (° C)98
  • Melting Point C (° C)-107
  • DescriptionIsooctane, OPTIMA™ Grade, Glass Bottle, 4 L
Documents & Certificates
Reviews
Q&A