Graphic Controls 00378661 Circular Chart, Mercury Instrument, 7 day
Representative image only.
Graphic Controls 00378661 Circular Chart, Mercury Instrument, 7 day
Graphic Controls | Mfr: 00378661| Item: UX-49000-62
Get exacting performance that meets American Gas Association (AGA) standards
$66.08USD / Box of 100
$94.40USD / Box of 100
$66.08USD / Box of 100
$94.40USD / Box of 100
Product Description
Key Features
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Optimal chart paper thickness prevents curling
  • Welker Slot designed for use with automatic chart changers
  • Scale reads 0 to 100
  • 1 year warranty
More About this Item
Use with Mercury chart recorders.
1 Year Warranty
Specifications
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)8
  • DescriptionCircular Chart, Mercury Instrument, 7 day