Reagents Titanium Dioxide, 500g, Purified

Representative image only.

Reagents Titanium Dioxide, 500g, Purified

Reagents | Mfr: C2340500-500A5| Item: UX-78914-52

Manufactured using the highest quality raw materials in ISO 9001 facility

$403.00USD / Each

$403.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • 20 years warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

20 Year Warranty
Specifications
  • Linear FormulaTiO2
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight79.87
  • PackagingPoly Jar
  • CAS Number13463-67-7
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • DescriptionTitanium Dioxide, 500g, Purified
  • Warranty20 years
Documents & Certificates
Reviews
Q&A