Spectrum Chemical Hydroxy Naphthol Blue Disodium Salt, Reagent ACS Grade; 25 g
仅限业务代表图片。
Spectrum Chemical Hydroxy Naphthol Blue Disodium Salt, Reagent ACS Grade; 25 g
Spectrum Chemical | Mfr: H1092-25GM| Item: EW-80100-97
Quality and delivery you can count on every time!
$119.50USD / 每个
$119.50USD / 每个
Product Description
主要特点
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years
关于该商品的更多信息
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
规格
  • 纯度等级ACS Reagent
  • 经验公式C20H12O11N2S3Na2
  • 线性公式C20H12O11N2S3Na2
  • 重量 (g)25
  • 分子量598.5
  • 包装Amber Glass
  • CAS 编号165660-27-5
  • 保质期 (天)730
  • 保质期 (月)24
  • 描述Hydroxy Naphthol Blue Disodium Salt, Reagent ACS Grade; 25 g
评论
常见问题与解答