Cole-Parmer® Disposable, Weighted Polyethylene, PVC, FEP Bailers, and Groundwater Filter
Large opening for easy tie-off, greater water flow, and improved sink rate
  • Individually sealed bailers are purified and rinsed with deionized water before packaging
  • All bailers are weighted for increased sink rate
These disposable bailers feature tapered ends and low-snag cord cleat to reduce hang-ups and snags. The angled nozzle ensures positive check-valve control. Choose from virgin polyethylene construction for strength and durability, PVC for strength and economy of polyethylene with transparent features, or FEP construction for high-purity applications. Double check-valve bailers allow you to sample from a specific depth (point-source sampling).
$50.55 - $907.20USD / Case of 24
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Number of Valves
Availability Pricing
COMPARE
12 Polyethylene 1
$327.60USD / Case of 24
COMPARE
12 Polyethylene 2
$327.60USD / Case of 24
COMPARE
36 Polyethylene 1
$281.40USD / Case of 24
COMPARE
36 Polyethylene 2
$339.70USD / Case of 24
COMPARE
48 Polyethylene 1
$409.50USD / Case of 24
COMPARE
48 Polyethylene 2
$427.65USD / Case of 24
COMPARE
36 FEP 1
$702.45USD / Case of 12
COMPARE
36 FEP 2
$907.20USD / Case of 12
COMPARE
36 Polyethylene 1
$255.70USD / Case of 24
COMPARE
36 Polyethylene 2
$322.35USD / Case of 24
COMPARE
36 FEP 1
$263.85USD / Case of 12
COMPARE
36 FEP 2
$380.65USD / Case of 12
COMPARE
36 Polyethylene 1
$292.70USD / Case of 24
COMPARE
36 Polyethylene 1
$351.25USD / Case of 24
COMPARE
12 CPVC 1
$304.80USD / Case of 24
COMPARE
36 CPVC 1
$316.35USD / Case of 24
COMPARE
48 CPVC 1
$409.50USD / Case of 24
COMPARE
Polyethersulfone
$50.55USD / Case of 1
Showing 1- 18 of 18