Concentrated Lab Clarifier
Keeps circulating baths clean and odor-free
  • Stainless steel compatible
  • Treats up to 200 gallons
This concentrated clarifier keeps your circulating bath water pure and odor free. One 8-oz (237-mL) bottle treats up to 200 gallons (757 L).
$55.60USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
PolyScience Polyclean Concentrated Clarifier, 8 oz (250 mL) EW-08796-00 Mfr # 004-300040
COMPARE
Concentrated Clarifier, 8 oz (250 mL)
$55.60USD / Each