Cole-Parmer® 96-Well DispoDialyzer
Save time — simultaneously prepare 96 samples from 25µl to 300µl
  • Over 95% sample recovery
  • Each well contains a separate membrane to eliminate sample contamination and leakage
  • Very high reproducibility between different wells
These 96-well dialyzers let you simultaneously prepare 96 samples. Use for applications such as salt removal, buffer exchange, parallel sample prep after fraction collection, detergent removal, HPLC, and more.
Each well contains an individual, high-quality, regenerated cellulose membrane that eliminates sample contamination and leakage. Membranes are free from sulfur and heavy metal contamination. Wells are sealed with 8-cap strips (included).
To use the dialyzer, simply place the sample into each well and assemble the strip caps. Float the entire plate in dialysis buffer. Then, retrieve the purified samples for downstream applications.
What’s included: twelve 8-cap strips to seal wells.
$2,820.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
COMPARE
10000
$2,820.00USD / Each