Sherwood Scientific Laboratory Fluid Bed Dryers
Rapidly remove moisture and solvents from samples
  • Ensures thorough drying
  • Stores a program with up to 16 steps
This compact benchtop unit uses a powerful air delivery system and heater to rapidly remove moisture or solvents from samples with up to 80% moisture content. The unit ensures thorough drying, even with fine or irregularly-shaped samples. Store a program with up to 16 steps to optimize airflow rates, temperature, and drying time.
For greater control, order the optional software and RS-232 cable to interface with a computer. The software allows you to program any number of steps, ending each step at a specific time, humidity, or outlet temperature.
$15,215.00 - $16,410.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Sample Volume Description
Availability Pricing
Sherwood Scientific 50100200 Programmable Fluid Bed Dryer, 110 VAC, 50/60 Hz EW-02658-40 Mfr # 50100202
COMPARE
110 2-L, 5-L, or 1-L (four 250 mL samples), depending on tub assembly used. Max weight 11 lbs (5 kg) Programmable Fluid Bed Dryer, 110 VAC, 50/60 Hz
$16,410.00USD / Each
Sherwood Scientific 50100201 Programmable Fluid Bed Dryer, 230 VAC, 50/60 Hz EW-02658-45 Mfr # 50100201
COMPARE
220 2-L, 5-L, or 1-L (four 250 mL samples), depending on tub assembly used. Max weight 11 lbs (5 kg) Programmable Fluid Bed Dryer, 230 VAC, 50/60 Hz
$15,215.00USD / Each