Thermo Scientific™ Nunc™ MicroWell™ 96-Well Non-Treated Microplates, Polypropylene
Low binding polypropylene is ideal for homogeneous assays and storage
  • Shared wall design increases the well volume
  • Nontreated polypropylene plates are solvent resistant and autoclavable
Achieve complete sample recovery with the Thermo Scientific™ Nunc™ MicroWell™ 96-well microplates. Plates are ideal for storage as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from −80° to 121°C (−112° to 250°F).
Choose from conical bottom wells for optimal sample recovery or round bottom wells which are wider for improved mixing.
$410.00 - $634.00USD / Case of 60
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sterile Well Shape Well Volume (mL)
Availability Pricing
Thermo Scientific Nunc 442587 Microplates, 96-Well, Polypropylene, Non-Treated, Non-Sterile, Conical, 0.3 mL; 60/Cs EW-01929-30 Mfr # 442587
COMPARE
No Conical 0.3
$410.00USD / Case of 60
Thermo Scientific Nunc 267245 Microplates, 96-Well, Polypropylene, Non-Treated, Non-Sterile, Round, 0.5 mL; 120/Cs EW-01929-32 Mfr # 267245
COMPARE
No Round 0.5
$578.00USD / Case of 120
Thermo Scientific Nunc 249944 Microplates, 96-Well, Polypropylene, Non-Treated, Non-Sterile, Conical, 0.45 mL; 120/Cs EW-01929-34 Mfr # 249944
COMPARE
No Conical 0.45
$576.00USD / Case of 120
Thermo Scientific Nunc 249946 Microplates, 96-Well, Polypropylene, Non-Treated, Sterile, Conical, 0.45 mL; 120/Cs EW-01929-38 Mfr # 249946
COMPARE
Yes Conical 0.45
$634.00USD / Case of 120