Oakton EcoTestr pH 2 Waterproof Pocket pH Tester

Representative image only.
Oakton Item # EW-35423-10
Easy-to-read display with an easy-to-afford price!
  • Ergonomically designed LCD means measurements can be seen at a glance
  • Economical and easy to use
  • Waterproof and dustproof housing designed for use in dirty and damp field conditions
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeTester
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.1
  • pH Accuracy±0.1 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Min Temperature (° C)-5
  • Max Temperature (° C)50
  • Min Temperature (° F)23
  • Max Temperature (° F)122
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery4 x A76 batteries (included)
  • Width (cm)4.1275
  • Height (cm)15.875
  • Width (in)1 5/8
  • Length (in)1 1/8
  • Height (in)6 1/4
  • DescriptionWaterproof Pocket pH Tester
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 67ISO 9001:2008 Certified Supplier

Key Features

  • Ergonomically designed LCD means measurements can be seen at a glance
  • Economical and easy to use
  • Waterproof and dustproof housing designed for use in dirty and damp field conditions
  • 1 year warranty

More About this Item

Simply dip and read-- the orientation of the display means no need to turn your head or tilt the tester. This low-cost meter is perfect for most applications. The single-junction electrode is recessed to protect against breakage. The unit offers up to three-point calibration with auto-buffer recognition for quick calibrations, automatic temperature compensation for accurate reading under changing conditions, hold function to lock measure value, and auto-off to prolong battery life. Built-in belt clip allows you to hang your meter when not in use.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat