Cole-Parmer® Kynar PVDF Gas Sampling Bags
Corrosive-resistant Kynar for all your application needs
  • Strong seams for added durability
All standard models have either an on/off valve (3⁄16'' nickel-plated brass hose barb) or a silicone septum in polypropylene housing; dual-valve models have both. Max temp: 160°F (71°C).
Standard Kynar Bags
Single-wide seam; available in 2-mil and 4-mil thick styles.
Dual-Valve Kynar Bags
Double-wide seamed bags have both on/off and septum valves for convenient mixing.
$56.60 - $162.50USD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Width (in) Valve Volume (Liters)
Availability Pricing
COMPARE
6 On/off 0.3
$67.30USD / Each
COMPARE
9 On/off 1.6
$75.00USD / Each
COMPARE
12 On/off 3.8
$82.60USD / Each
COMPARE
15 On/off 8.1
$96.20USD / Each
COMPARE
18 On/off 20.3
$96.10USD / Each
COMPARE
24 On/off 37.7
$114.50USD / Each
COMPARE
30 On/off 85.7
$146.25USD / Each
COMPARE
6 Septum 0.3
$56.80USD / Each
COMPARE
9 Septum 1.6
$56.60USD / Each
COMPARE
12 Septum 3.8
$67.00USD / Each
COMPARE
18 Septum 20.3
$86.50USD / Each
COMPARE
24 Septum 37.7
$105.50USD / Each
COMPARE
30 Septum 85.7
$137.25USD / Each
COMPARE
6 Dual valve, on/off and septum 0.3
$79.80USD / Each
COMPARE
9 Dual valve, on/off and septum 1.6
$80.90USD / Each
COMPARE
12 Dual valve, on/off and septum 3.8
$89.80USD / Each
COMPARE
15 Dual valve, on/off and septum 8.1
$113.75USD / Each
COMPARE
18 Dual valve, on/off and septum 20.3
$111.75USD / Each
COMPARE
24 Dual valve, on/off and septum 37.7
$129.00USD / Each
COMPARE
30 Dual valve, on/off and septum 85.7
$162.50USD / Each
Showing 1- 20 of 20