Secchi Disk
Use to determine turbidity or degree of visibility in natural water supplies
  • Braided, stretch resistant line marked at half and full meter intervals up to 20 meters
Simple-to-use secchi disk is a weighted, 20-cm dia black and white plate that sinks parallel to the water's surface. For best results use, from the leeward side of a boat or dock between mid-morning and mid-afternoon.
$161.40USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Calibration Temperature compensation Waterproof
Availability Pricing
LaMotte 0171-CL Secchi Turbidity Disk, 20 m cable EW-05492-00 Mfr # 0171-CL
COMPARE
$161.40USD / Each