Environmental Express Oakton pH 5+ Handheld Meter with ATC; Order pH Probe Separately

Representative image only.

Environmental Express Oakton pH 5+ Handheld Meter with ATC; Order pH Probe Separately

Environmental Express Oakton | Item: EW-35613-50

High +/-0.01 pH accuracy at an economical price

$478.00USD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$478.00USD / Each
Product Description

Includes

4 AAA batteries.

Key Features

  • Convenient, compact design
  • Up to five-point push-button calibration with autobuffer recognition
  • Displays pH and temperature with the press of a button
  • 3 year warranty

More About this Item

These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 80°C. Meters include a protective rubber boot that also acts as a meter stand.

Meter only; order probe separately. 59001-70 Double Junction pH electrode, 59001-65 Single Junction pH electrode


What's Included: ATC probe, protective rubber boot, and batteries.

3 Year WarrantyConformite Europeenne
Specifications
  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Temperature compensationAutomatic
  • Battery4 AAA (included)
  • Battery life (hr)700
  • Width (cm)8.5725
  • Height (cm)4.28625
  • Width (in)3 3/8
  • Height (in)1 11/16
  • DescriptionHandheld Meter with ATC; Order pH probe separately
  • Warranty3 year
Reviews
Q&A