A&D Weighing TB-220PA Type C Plug Adapter for Continental European Countries
Representative image only.
A&D Weighing TB-220PA Type C Plug Adapter for Continental European Countries
A&D Weighing | Mfr: TB-220PA| Item: UX-11112-52
$4.50USD / Each
$5.00USD / Each
for an additional
$206.50USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$4.50USD / Each
$5.00USD / Each
Buy It Together
Current Item
A&D Weighing TB-220PA Type C Plug Adapter for Continental European Countries
A&D Weighing TB-220PA Type C Plug Adapter for Continental European Countries
1111252
$4.50USD / Each
$5.00USD / Each
+ + + + +
Environmental Express Buffer Solution, pH 4.01; 500 mL
0065400
$20.10USD / Pint of 1
+ + + +
Environmental Express Electrode Storage Solution; 500 mL
0065304
$27.30USD / Pint of 1
=
Total Price 2751.80 ADD 10 ITEMS TO CART
Product Description
Key Features
  • 2 year warranty
More About this Item
Choose the adapter to meet your power needs.
Mechanical adapter to transition from North American style blade prongs to Type C round prongs found in most continental European countries and others with 220V mains
Two year warranty
Specifications
  • Power (VAC)230
  • DescriptionType C Plug Adapter for Continental European Countries
  • Warranty2 Years