Ace Glass 5028-24 Tube Adapter, 10/18 Joint
Representative image only.
Ace Glass 5028-24 Tube Adapter, 10/18 Joint
Ace Glass | Mfr: 5028-24| Item: UX-34721-00
$60.00USD / Each
$60.00USD / Each
Product Description
Key Features
  • Adapter has a Standard Taper bottom joint and #7 Ace threads on top.
More About this Item
Nylon bushing tightens on top threads to form an O-ring compression seal for use with thermometers and bleed tubes up to 7 mm in diameter. All adapters include nylon bushing and FETFE O-ring.