Acros Organics Benzalkonium chloride, 50 wt% aqueous solution (1l)

Representative image only.

Acros Organics Benzalkonium chloride, 50 wt% aqueous solution (1l)

Acros Organics | Mfr: AC263820010| Item: UX-88256-50

Quality and consistency your lab can count on

$430.00USD / Each

$430.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Corrosive
Specifications
  • Linear FormulaC6H5CH2N(CH3)2RCl R=C8H 17 to C 18 H 37
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • SynonymsAlkyl-benzyl-dimethylammonium chloride
  • Volume (mL)1000
  • PackagingGlass bottle
  • DOT InformationCORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.; Class 8, Packing Group II
  • Hazard SymbolCorrosive
  • UN NumberUN3265
  • CAS Number8001-54-5
  • DescriptionBenzalkonium chloride, 50 wt% aqueous solution (1l)
Documents & Certificates
Reviews
Q&A